Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEI3 Rabbit anti-Human, Alexa Fluor 488, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100441AF488
Description
NEI3 Polyclonal antibody specifically detects NEI3 in Human samples. It is validated for Western BlotSpecifications
NEI3 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 488 | |
DNA glycosylase FPG2, DNA glycosylase hFPG2, DNA glycosylase/AP lyase Neil3, EC 3.2.2.-, EC 4.2.99.18, endonuclease 8-like 3, Endonuclease VIII-like 3, FGP2, FLJ10858, FPG2, hFPG2, hNEI3, nei endonuclease VIII-like 3 (E. coli), NEI3, Nei-like protein 3 | |
Residues 164-177 (LRAESEVKKQKGRMLG) of the human NEI3 protein. 85% sequence identity to mouse and rat. | |
0.1 mL | |
DNA Repair | |
55247 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction