Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK10 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP31013925UL

 View more versions of this product

Catalog No. NB125180


Add to cart

Description

Description

NEK10 Polyclonal antibody specifically detects NEK10 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

NEK10
Unconjugated
PBS buffer, 2% sucrose
FLJ32685, NIMA (never in mitosis gene a)- related kinase 10
The immunogen is a synthetic peptide directed towards the middle region of mouse NEK10 (NP_001182158.1). Peptide sequence HLGSGAFGCVYKVRKRSGQNLLAMKEVNLHNPAFGKDKKDRDSSVKNIVS
25 μg
Protein Kinase
Primary
152110
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit