Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK11 Mouse anti-Human, FITC, Clone: OTI4H5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272933F
Description
NEK11 Monoclonal specifically detects NEK11 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
NEK11 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
NEK11 | |
Full length recombinant protein of human NEK11 (NP_079076) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunocytochemistry, Immunofluorescence | |
OTI4H5 | |
Immunocytochemistry/Immunofluorescence | |
EC 2.7.11, EC 2.7.11.1, FLJ23495, Never in mitosis A-related kinase 11, NIMA (never in mitosis gene a)- related kinase 11, NimA-related protein kinase 11, serine/threonine-protein kinase Nek11 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
79858.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title