Learn More
Aviva Systems Biology Corp NEK3 Antibody - N-terminal region (ARP49070_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49070_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NEK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NEK3. Alternate names: HSPK36, MGC29949ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60010