Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK3 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548646

Catalog No. PA548646

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with pig and rat based on sequence homology.

This gene encodes a member of the NimA (never in mitosis A) family of serine/threonine protein kinases. The encoded protein differs from other NimA family members in that it is not cell cycle regulated and is found primarily in the cytoplasm. The kinase is activated by prolactin stimulation, leading to phosphorylation of VAV2 guanine nucleotide exchange factor, paxillin, and activation of the RAC1 GTPase. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

Nek3
Polyclonal
PBS with 0.09% sodium azide
Q9R0A5
Serine/threonine-protein kinase Nek3, Never in mitosis A-related kinase 3, NimA-related protein kinase 3, Nek3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23954
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Nek3
Liquid
NEK3
KLH conjugated synthetic peptide between 446-474 amino acids from the C-terminal region of mouse Nek3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit