Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK5 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548697

Catalog No. PA548697

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Nek5
Polyclonal
PBS with 0.09% sodium azide
Q7TSC3
Serine/threonine-protein kinase Nek5, Never in mitosis A-related kinase 5, NimA-related protein kinase 5, Nek5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
330721
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Nek5
Liquid
NEK5
KLH conjugated synthetic peptide between 525-552 amino acids from the C-terminal region of mouse Nek5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit