Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK6 Mouse anti-Human, Mouse, Rat, FITC, Clone: OTI5D7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272938F
Description
NEK6 Monoclonal specifically detects NEK6 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
NEK6 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
NEK6 | |
Full length recombinant protein of human NEK6 (NP_055212) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5D7 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 2.7.11.1, Never in mitosis A-related kinase 6, NIMA (never in mitosis gene a)-related kinase 6, NimA-related protein kinase 6, Protein kinase SID6-1512, putative serine-threonine protein kinase, serine/threonine-protein kinase Nek6, SID6-1512 | |
Mouse | |
Affinity Purified | |
GPCR, Protein Kinase | |
10783.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title