Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK6 Mouse anti-Human, Clone: OTI5B9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524947

Catalog No. PIMA524947


Add to cart

Description

Description

The Aspergillus nidulans  ft.never in mitosis A ft. (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA. NEK6 encodes a NEK type protein kinase, which contains a C-terminal NIMA-like catalytic domain.
Specifications

Specifications

NEK6
Monoclonal
1.5 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HC98
Serine/threonine-protein kinase Nek6; Never in mitosis A-related kinase 6; NimA-related protein kinase 6; Nek6
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10783
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
OTI5B9
Unconjugated
NEK6
Liquid
NEK6
Full length human recombit protein of NEK6 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit