Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NEK7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551334
Description
The antibody detects endogenous levels of total NEK7 protein.
NEK7 (NIMA Related Kinase 7) is a Protein Coding gene. Diseases associated with NEK7 include Muckle-Wells Syndrome and Cinca Syndrome. Among its related pathways are Cell Cycle, Mitotic and Transport of the SLBP independent Mature mRNA. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is NEK6.
Specifications
NEK7 | |
Polyclonal | |
Unconjugated | |
NEK7 | |
2810460C19Rik; AU020186; NEK7; Never in mitosis A-related kinase 7; NIMA (never in mitosis gene a)-related expressed kinase 7; NIMA (never in mitosis gene a)-related kinase 7; NIMA related kinase 7; NIMA-related expressed kinase 7; NIMA-related kinase 7; nimA-related protein kinase 7; Serine/threonine-protein kinase Nek7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140609, 360850, 59125 | |
-20°C | |
Liquid |
Western Blot | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
D3ZBE5, Q8TDX7, Q9ES74 | |
NEK7 | |
Synthetic peptide of human NEK7. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction