Learn More
Sklar InstrumentsSupplier Diversity Partner Nelaton Grooved Director, Sklar, 6-1/4 in., Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 301210
Nelaton Grooved Director, Sklar, 6-1/4 in., Stainless Steel. Sklar's Nelaton grooved director is commonly used in examining the mouth of a pediatric patient. It is also used in the treatment of ankyloglossia, or in.tongue tie. in. One end of the instrument is flat and pear-shaped. The distal end split assists in isolating the tongue and lingual frenium. The probe end is used to examine the rest of the mouth.
Catalog No. 50-143-8103