Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neo/GUIDE Microscope Slide Sets
Specifications
Slide Type | Prepared |
---|
Includes: all slides from the following sets described above: Mitosis and Meiosis, Five Kingdoms, Survey of Algae, Plant Anatomy, Survey of Animals, and Human Body.
Catalog Number | Mfr. No. | For Use With (Application) | Quantity | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Quantity | Type | Price | Quantity & Availability | |||
S67710
![]() |
Neo/Sci Corporation
704006 |
View the three forms of bacteria on one slide | 1 slide | Bacteria Structure Slide Set |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S67728
![]() |
Neo/Sci Corporation
704016 |
A microscopic investigation into monera, protists, fungi, plants, and animals kingdoms | Set of 20 slides | 5 Kingdoms Slide Set |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S67724
![]() |
Neo/Sci Corporation
704014 |
Microscopic investingation into various organism | Set of 41 slides | Exploring Life I Slide Set |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S67715
![]() |
Neo/Sci Corporation
704205 |
A close-up view of the microworld | Class set: 5 sets of 5 slides | Exploring Microlife Slide Set |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S67716
![]() |
Neo/Sci Corporation
704007 |
A close-up view of the microworld | Set of 5 slides | Exploring Protists Slide Set |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S67706
![]() |
Neo/Sci Corporation
704003 |
Investigate the microworld of common things | Set of 5 slides | Exploring Common Things Slide Set |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S67709
![]() |
Neo/Sci Corporation
704204 |
Investigate the microscopic world of cells | Class set: 5 sets of 5 slides | Exploring Cells Slide Set |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S67726
![]() |
Neo/Sci Corporation
704017 |
Microscopic investigation into cells division and reproduction | Set of 5 slides | Mitosis and Meiosis Slide Set |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S67737
![]() |
Neo/Sci Corporation
704218 |
A microscopic look at the systems of the human body: squamous epithelium, adipose tissue, hyaline cartilage, striated muscle and compact bone. | 5/pk. | Humany Body Systems |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S67714
![]() |
Neo/Sci Corporation
704005 |
A close-up view of the microworld | Set of 5 slides | Exploring Microlife Slide Set |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S67727
![]() |
Neo/Sci Corporation
704217 |
Microscopic investigation into cells division and reproduction | 5/pk. | Mitosis and Meiosis Slide Set |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Sets come complete with Neo/GUIDE reference booklet featuring detailed background information and structure identification key. Available as single sets or classroom packs of multiple sets.Common Things (Set of 5 slides)
- Feather (three types)
- Housefly head, leg, and wing
- Textile fibers
- Hair (three types)
- Letter “e”
- Bacteria (Prokaryotic cell)
- Cheek (Eukaryotic cell)
- Cyanobacteria (Photosynthetic bacteria)
- Mixed protozoa
- Lilac leaf (plant cell)
- Bacteria
- Bread mold
- Yeast
- Planktonic forms
- Mixed protozoa
- Contains smears of cocci, bacilli and spirilla on one slide
- Paramecium
- Amoeba
- Spirogyra
- Mixed protists
- Euglena
- Onion mitosis
- Plant root
- Plant stem
- Plant leaf
- Plant flower bud
- Daphnia
- Planaria
- Hydra
- Plankton
- Rotifers
- Trichinella (Trichinosis)
- Plasmodium (Malaria)
- Pneumocystis (AIDS-opportunistic infection)
- Mycrobacterium (Tuberculosis)
- Entamoeba (Amoebic dysentery)
- Aspergillus
- Molds (three types)
- Mushroom
- Bread mold
- Sporigia and zygotes
- Budding yeast
Includes all slides from Common Things, Cells, Microlife, Bacteria, Protists, Plants, Invertebrates, Disease-causing Organisms, and Fungi Sets, described above.
Survey of Algae (Set of 10 slides)
- Euglena, wm
- Volvox, wm
- Chlamydomonas, wm
- Spirogyraconjugation, wm
- Oedogonium, wm
- Fucus, cs
- Porphyra, wm
- MoneraMixed bacteria, Anabaena, Oscillatoria
- ProtistaVorticella, Paramecium, Amoeba, Volvox
- FungiPenicillium, Rhizopus Stolonifer
- PlantaeMoss leaf, Pinus leaf, Fern (Prothallium), Helianthus stem
- AnimaliaHydra, Planaria, Bed bug, Earthworm, Sea urchin, Fish scales, Frog skin
- Onion root tip mitosis
- Whitefish blastula mitosis
- Meiosis I in plant cells
- Meiosis II in plant cells
- Meiosis in animal cells (Grasshopper Testis)
- TissuesSquamous Epithelium, Adipose tissue, Striated muscle
- SkeletalCompact bone, Hyaline cartilage
- NervousNerve cells, Spinal cord, Cerebellum
- CirculatoryArtery and Vein, Blood smear (Giemsa stain)
- OrgansLiver, Stomach, Small intestine, Lung, Spleen, Colon, Thyroid Gland, Pancreas, Human skin, Kidney
- Monocot and dicot root
- Monocot and dicot stem
- Monocot and dicot leaf
- Monocot and dicot flower bud
- Deciduous and evergreen leaf
- Grantia
- Hydra
- Planaria
- Liver fluke
- Taenia pisiformis
- Ascaris
- Earthworm
- ;Daphnia
- Spider (Head and mouthparts)
- Mites
- Coackroach (Head and mouthparts)
- Locust (Mouthparts)
- Locust (Thorax)
- Locust (Abdomen)
- Drosophila (Wild type)
- Culex (Pupa)
- Aphid
- Sea urchin (Ova)
- Fish scales
- Frog skin
Specifications
Prepared |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title