Alfa Aesar™ Neodymium boride, 99.5%

Manufacturer: Alfa Aesar™ 3949414

Quantity 25g

 View more versions of this product

Catalog No. AA3949414

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 12008-23-0
    Formula Weight 209.1
    MDL Number MFCD00151359
    Chemical Name or Material Neodymium boride

    Specifications

    Formula Weight 209.1
    Odor Odorless
    CAS 12008-23-0
    TSCA TSCA
    Mesh Size −325 Mesh
    MDL Number MFCD00151359
    Quantity 25g
    Chemical Name or Material Neodymium boride
    Assay 99.5%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit