Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Neodymium foil, 0.1mm (0.004 in.) thick, 99.9% (REO)

Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
7440-00-8
Nd
144.242
MFCD00011130
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
23934
CHEBI:33372
neodymium
[Nd]
7440-00-8
Nd
144.242
MFCD00011130
QEFYFXOXNSNQGX-UHFFFAOYSA-N
neodimio, neodyme, neodym, trihydride, ingot reo, neodymium, ion nd5+, atom, ingot, iii, foil, 3n
23934
CHEBI:33372
neodymium
[Nd]
  • Packaged under argon
Weight ≈0.43g/25x25mm
Quantity 25 x 25 mm
UN Number UN3178
Solubility Information Reacts with water.
Odor Odorless
Packaging Mylar under Argon
Physical Form Foil
Percent Purity 99.9%
Chemical Name or Material Neodymium foil, 0.1mm (0.004 in.) thick
Assay (REO)

GHS H Statement
H260-H228
In contact with water releases flammable gases which may ignite spontaneously.
Flammable solid.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-109-3

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures