Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Neodymium(III) 2,4-pentanedionate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1318409

Quantity 10g

 View more versions of this product

Catalog No. AA1318409


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

14589-38-9
MFCD00013498
Neodymium(III) 2,4-pentanedionate
441.57
Neodymium acetylacetonate

Specifications

14589-38-9
441.57
Neodymium acetylacetonate
150° to 152°C
Neodymium(III) 2,4-pentanedionate
Powder
MFCD00013498
Odorless
10g
99.9%, (REO)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit