Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) 2,4-pentanedionate, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number AA1318404
Click to view available options
Quantity:
2 g
10 g
14589-38-9
C15H21NdO6
441.57
MFCD00013498
AZTDRGMPTCCVFW-UHFFFAOYSA-N
Neodymium acetylacetonate
122177161
[Nd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
14589-38-9
C15H21NdO6
441.57
MFCD00013498
AZTDRGMPTCCVFW-UHFFFAOYSA-N
Neodymium acetylacetonate
122177161
[Nd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 150°C to 152°C
Physical Form Powder
Quantity 2 g
IUPAC Name neodymium(3+) tris(2,4-dioxopentan-3-ide)
Formula Weight 441.57
Percent Purity 99.9%
Odor Odorless
Assay (REO)
Chemical Name or Material Neodymium(III) 2,4-pentanedionate
EINECSNumber 238-631-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit