Alfa Aesar™ Neodymium(III) carbonate hydrate, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1530118

Quantity 50g

 View more versions of this product

Catalog No. AA1530118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38245-38-4
    Formula Weight 468.51 (Anhydrous)
    MDL Number MFCD00150426
    Chemical Name or Material Neodymium(III) carbonate hydrate

    Specifications

    Formula Weight 468.51 (Anhydrous)
    Physical Form Powder
    MDL Number MFCD00150426
    Quantity 50g
    TSCA TSCA
    CAS 38245-38-4
    Chemical Name or Material Neodymium(III) carbonate hydrate
    Assay 99.9%, (REO)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit