Neodymium(III) chloride hydrate, 99.9% (REO), Alfa Aesar™ Reacton™

Manufacturer: Alfa Aesar 1125118

Quantity 50g

 View more versions of this product

Catalog No. AA1125118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10024-93-8
    Physical Form Crystalline Aggregates
    Assay Percent Range 99.9% (REO)
    Molecular Formula NdCl3·H2O
    MDL Number MFCD00011132
    Chemical Name or Material Neodymium(III) chloride hydrate

    Specifications

    Physical Form Crystalline Aggregates
    Quantity 50g
    EINECS Number 233-031-5
    Odor Odorless
    Assay Percent Range 99.9% (REO)
    Molecular Formula NdCl3·H2O
    Appearance Crystalline aggregrates
    MDL Number MFCD00011132
    CAS 10024-93-8
    Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Neodymium(III) chloride hydrate

    H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit