Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) fluoride, 98%, Acros Organics

Click to view available options
Quantity:
10g
50g
Packaging:
Plastic Bottle
13709-42-7
F3Nd
MFCD00011135
13709-42-7
F3Nd
MFCD00011135
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula NdF3
Formula Weight 201.23
Physical Form Powder
Percent Purity 98%
Packaging Plastic Bottle
Solubility Solubility in water: insoluble.
Boiling Point 2300.0°C
Color Gray
Melting Point 1410.0°C
Quantity 10g
Chemical Name or Material Neodymium(III) fluoride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with acids liberates very toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger