Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Neodymium(III) nitrate hexahydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1291230

Quantity 250g

 View more versions of this product

Catalog No. AA1291230


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

16454-60-7
MFCD00149799
14,6450
438.34 (330.25 Anhydrous)
UN1477
Neodymium(III) nitrate hexahydrate

Specifications

16454-60-7
438.34 (330.25 Anhydrous)
UN1477
Odorless
≈40°C
250g
99.9%, (REO)
Crystalline
MFCD00149799
14,6450
Hygroscopic
3.0 to 4.0
Neodymium(III) nitrate hexahydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

RTECSNumber : OJ6125000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit