Alfa Aesar™ Neodymium(III) oxide, REacton™, 99.997% (REO)

Manufacturer: Alfa Aesar™ 1383722

Quantity 100g

 View more versions of this product

Catalog No. AA1383722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1313-97-9
    Formula Weight 336.48
    MDL Number MFCD00011134
    Merck Index 14,6450
    Chemical Name or Material Neodymium(III) oxide
    Grade Reacton™

    Specifications

    Formula Weight 336.48
    Physical Form Powder
    MDL Number MFCD00011134
    Merck Index 14,6450
    Melting Point 2272°C
    Quantity 100g
    Density 7.24g/mL
    TSCA TSCA
    CAS 1313-97-9
    Chemical Name or Material Neodymium(III) oxide
    Assay 99.997%, (REO)
    Grade Reacton™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit