Alfa Aesar™ Neodymium(III) oxide, REacton™, 99.999% (REO)

Manufacturer: Alfa Aesar™ 4556614

Quantity 25g

 View more versions of this product

Catalog No. AA4556614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1313-97-9
    Molecular Formula Nd2O3
    Formula Weight 336.48
    MDL Number MFCD00011134
    Merck Index 14,6450
    Chemical Name or Material Neodymium (III) oxide
    Grade Reacton™

    Specifications

    Formula Weight 336.48
    TSCA TSCA
    MDL Number MFCD00011134
    Density 7.24g/mL
    Physical Form Powder
    Molecular Formula Nd2O3
    CAS 1313-97-9
    Merck Index 14,6450
    Color Pale blue
    Melting Point 2272°C
    Quantity 25g
    Chemical Name or Material Neodymium (III) oxide
    Assay 99.999%, (REO)
    Grade Reacton™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit