Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) oxide, REacton™, 99% (REO), Thermo Scientific Chemicals

Catalog Number aa4025214
Click to view available options
Quantity:
25 g
100 g
500 g
1313-97-9
Nd2O3
336.48
MFCD00011134
PLDDOISOJJCEMH-UHFFFAOYSA-N
dineodymium(3+) trioxidandiide
[O--].[O--].[O--].[Nd+3].[Nd+3]
1313-97-9
Nd2O3
336.48
MFCD00011134
PLDDOISOJJCEMH-UHFFFAOYSA-N
dineodymium(3+) trioxidandiide
[O--].[O--].[O--].[Nd+3].[Nd+3]

ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst.Neodymium(III) oxide is an inorganic compound containing inner transition metal neodymium from lanthanides series in +3 oxidation state and oxide in 2:3 ratio. It adopts a hexagonal crystal structure. It is also known as neodymium oxide, neodymiumtrioxide, neodymium(3+)oxide, neodymium sesquioxide and dineodymiumtrioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst
TRUSTED_SUSTAINABILITY
Melting Point 2272°C
Physical Form Powder
Quantity 25 g
Merck Index 14,6450
Solubility Information Insoluble in water; Soluble in acids.
Formula Weight 336.48
Percent Purity 99%
Grade Reacton™
Density 7.24 g/mL
Assay (REO)
Chemical Name or Material Neodymium(III) oxide
Show More Show Less
EINECSNumber 215-214-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit