Learn More
Neodymium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals
Description
Neodymium(III) perchlorate used in the preparation of complexes of phosphine oxides. Two three-dimensional frameworks composed of doubly interpenetrating networks of the same topology but different stereoisomeric structures have been constructed from self-assembly of the chiral building block, Δ- or Λ-[Nd(ntb)2]3+.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Liquid |
Quantity | 25 g |
UN Number | UN3211 |
Solubility Information | Insoluble in water. |
Formula Weight | 550.68 (442.59 Anhydrous) |
Grade | Reagent |
Concentration or Composition (by Analyte or Components) | 50% w/w aq. soln. |
Odor | Odorless |
Chemical Name or Material | Neodymium(III) perchlorate |
Safety and Handling
Hazard Category | H271-H315-H319-H335 |
Hazard Statement | GHS H Statement H272-H315-H319-H335 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S. |
EINECSNumber | 236-821-8 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.