Alfa Aesar™ Neodymium powder, -200 mesh, 99.9% (REO)

Manufacturer: Alfa Aesar™ 4591414

Quantity 25g

 View more versions of this product

Catalog No. AA4591414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-00-8
    MDL Number MFCD00011130
    UN Number UN1383
    Chemical Name or Material Neodymium powder

    Specifications

    Quantity 25g
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P222-P231+P232-P370+P378c-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    Handle under inert gas.
    Protect from moisture.
    In case of fire:
    Color gray to silver
    TSCA TSCA
    Physical Form Powder
    Health Hazard 2 GHS H Statement
    H250-H260-H252
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.
    Mesh Size −200 Mesh
    Sensitivity Air & Moisture Sensitive
    Odor Odorless
    MDL Number MFCD00011130
    CAS 7440-00-8
    Storage Note 1
    • Packaged under Argon
    RTECS Number QO8575000
    UN Number UN1383
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC METALS, N.O.S.
    Chemical Name or Material Neodymium powder
    Assay 99.9%, (REO)

    GHS H Statement
    H250-H260-H252
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.

    GHS P Statement
    P210-P222-P231+P232-P370+P378c-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    Handle under inert gas.
    Protect from moisture.
    In case of fire:

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit