Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Neodymium powder, -325 mesh, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 45239-22

Quantity 100g

 View more versions of this product

Catalog No. AA4523922

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7440-00-8
MFCD00011130
UN1383
Neodymium powder

Specifications

7440-00-8
MFCD00011130
Silver
Odorless
Neodymium powder
Powder
UN1383
100g
−325 Mesh
99.9%, (REO)

Safety and Handling

GHS H Statement
H250-H260-H252

GHS P Statement
P210-P222-P231+P232-P280-P235+P410-P233-P422a-P420-P407-P501a

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC METALS, N.O.S.

RTECSNumber : QO8575000

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit