Alfa Aesar™ Neodymium powder, -325 mesh, 99.9% (REO)

Manufacturer: Alfa Aesar™ 4523914

Quantity 25g

 View more versions of this product

Catalog No. AA4523914

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7440-00-8
    MDL Number MFCD00011130
    UN Number UN1383
    Chemical Name or Material Neodymium powder

    Specifications

    Quantity 25g
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD00011130
    UN Number UN1383
    Health Hazard 2 GHS H Statement
    H250-H260-H252
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.
    Health Hazard 3 GHS P Statement
    P210-P222-P231+P232-P370+P378c-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    Handle under inert gas.
    Protect from moisture.
    In case of fire:
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC METALS, N.O.S.
    Mesh Size −325 Mesh
    TSCA TSCA
    CAS 7440-00-8
    Health Hazard 1 Danger
    RTECS Number QO8575000
    Color Silver
    Chemical Name or Material Neodymium powder
    Assay 99.9%, (REO)

    GHS H Statement
    H250-H260-H252
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.

    GHS P Statement
    P210-P222-P231+P232-P370+P378c-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    Handle under inert gas.
    Protect from moisture.
    In case of fire:

    • Packaged under Argon

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit