Alfa Aesar™ Neodymium rod, 6.35mm (0.25 in.) dia., 99.5% (metals basis excluding Ta)

Manufacturer: Alfa Aesar™ 44051DJ

Quantity 50mm

 View more versions of this product

Catalog No. AA44051DJ

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-00-8
    MDL Number MFCD00011130
    Chemical Name or Material Neodymium rod, ≈6.35mm (0.25 in.) dia.

    Specifications

    Physical Form Cast Rod
    Quantity 50mm
    Packaging Mylar under Argon
    Weight ≈2.2g/mm
    MDL Number MFCD00011130
    CAS 7440-00-8
    Odor Odorless
    TSCA TSCA
    Storage Note 1
    • Packaged in mylar under argon
    RTECS Number QO8575000
    Chemical Name or Material Neodymium rod, ≈6.35mm (0.25 in.) dia.
    Assay 99.5%, (metals basis excluding Ta)

    • Packaged in mylar under argon

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit