Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium standard solution, for AAS, 1mg/mL Nd in 2% HNO3, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
7440-00-8,7697-37-2,7732-18-5
Nd
144.242
MFCD00011130
QEFYFXOXNSNQGX-UHFFFAOYSA-N
23934
CHEBI:33372
neodymium
[Nd]
7440-00-8,7697-37-2,7732-18-5
Nd
144.242
MFCD00011130
QEFYFXOXNSNQGX-UHFFFAOYSA-N
23934
CHEBI:33372
neodymium
[Nd]
for atomic adsorption spectroscopy
CAS Min % 1.9,96.9
CAS Max % 2.1,97.1
Concentration 995 to 1005 μg/mL of Nd in 2% HNO3 (20°C)
Formula Weight 144.24
Physical Form Solution
Grade AAS
Packaging Plastic bottle
Color Undesignated
Quantity 100mL
Chemical Name or Material Neodymium standard solution, 1 mg/ml Nd in 2% HNO3

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning