Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ Neonatal 17OHP Coated Tube RIA Kit

The Neonatal 17OHP Coated Tube RIA Kit is for newborn screening.

Manufacturer:  MP Biomedical™ 07271305

 View more versions of this product

Catalog No. MP07271305

This product is regulated by the federal government, and may not be ordered online.


Description

Description

  • The Neonatal 17OHP Coated Tube RIA Kit is designed for the quantitative determination of 17OHP in neonatal blood collected on filter paper.
  • Sample Type and Volume: Dried Blood Spot, 3 mm (1/8 in.)
Specifications

Specifications

Newborn Screening
Blood, 3 mm (1/8 in.)
500 Tests
Blood
FDA cleared for In Vitro Diagnostic (IVD) use in the United States. CE Marked for IVD use outside the United States.
17OHP (17 alpha hydroxyprogesterone)
Human
Kit and Assay
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

FDA cleared for In Vitro Diagnostic (IVD) use in the United States. CE Marked for IVD use outside the United States.