Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neopentyliodide, stabilized, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
15501-33-4
C5H11I
198.047
MFCD00001083
CJTZXIJETZZARD-UHFFFAOYSA-N
neopentyl iodide,propane, 1-iodo-2,2-dimethyl,2,2-dimethyl-1-iodopropane,neopentyliodide,2,2-dimethylpropyl iodide,acmc-20ak50,1-iodo-2,2-dimethyl-propane,propane,1-iodo-2,2-dimethyl,1-iodo-2,2-dimethylpropane,1-iodo-2,2-dimethylpropane, technical gc
84953
1-iodo-2,2-dimethylpropane
CC(C)(C)CI
15501-33-4
C5H11I
198.047
MFCD00001083
CJTZXIJETZZARD-UHFFFAOYSA-N
neopentyl iodide,propane, 1-iodo-2,2-dimethyl,2,2-dimethyl-1-iodopropane,neopentyliodide,2,2-dimethylpropyl iodide,acmc-20ak50,1-iodo-2,2-dimethyl-propane,propane,1-iodo-2,2-dimethyl,1-iodo-2,2-dimethylpropane,1-iodo-2,2-dimethylpropane, technical gc
84953
1-iodo-2,2-dimethylpropane
CC(C)(C)CI
Stabilizer copper
Refractive Index 1.4880 to 1.4900 (20°C, 589nm)
Density 1.493g/mL
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3)3CCH2I
Formula Weight 198.04
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Neopentyl iodide
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Product may discolor during storage