Learn More
United Scientific Supplies Neoprene Stoppers Products for Science Education
Used in areas where rubber would be attacked by reagents or solvents. United Scientific Supplies Neoprene Stoppers have good resiliency and shelf-life.
Manufacturer:  United Scientific Supplies NST5AS
 View more versions of this product
Catalog No. S23957
Add to cart
Description
Description
  • Withstand petroleum products and oils as well as most inorganic acids and bases
  • Green for easy identification
  • Sold by the pound
Specifications
Specifications
Approx. No. per lb.: 21
Neoprene
Stopper
Green
Education