Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Neoprene Stoppers
Used in areas where rubber would be attacked by reagents or solvents. Neoprene Stoppers have good resiliency and shelf-life.
Supplier: United Scientific™ NST6S
Alert:
Prop 65 Warning
Description
- Withstand petroleum products and oils as well as most inorganic acids and bases.
- Green for easy identification.
- Sold by the pound.
Warranty and Services
1 Year
Specifications
Stopper | |
Tapered | |
Autoclavable | |
6 | |
Approx. No. per lb.: 19 |
26 mm | |
Non-sterile | |
32 mm | |
Neoprene | |
Green |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction