NEOS Voyager™ STABILicers™ Overshoes

The ultimate traction in snow and ice

Manufacturer: Honeywell Safety Products VNS1BLKXSM

 View more versions of this product

Catalog No. 19-920-162

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ANSI Z41-1991, OSHA 1910.136
    Closure Type Hook and loop strap
    Cuff Style Hook and Loop closure
    For Use With (Application) Agriculture, construction, forestry, municipal services, transportation, utilities, wind energy
    Gender Male
    Height (English) 11 in.
    Sole Features High traction Rubber outsole with replaceable steel cleats - cleat wrench included with every pair
    Waterproof Yes
    Color Black
    Material Nylon
    Size X-Small

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit