Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine, 98%

Click to view available options
Quantity:
250mg
1g
5g
148083-64-1
C27H26N4O8
534.525
MFCD00273454
OENCMORJQAUAAJ-DEOSSOPVSA-N
fmoc-lys dnp-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-2,4-dinitrophenyl amino hexanoic acid,fmoc-l-lys dnp-oh,n-fmoc-n'-2,4-dinitrophenyl-l-lysine,fmoc-lys dnp,2s-6-2,4-dinitrophenyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-2,4-dinitrophenyl-l-lysine
44593781
(2S)-6-(2,4-dinitroanilino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCNC4=C(C=C(C=C4)[N+](=O)[O-])[N+](=O)[O-])C(=O)O
148083-64-1
C27H26N4O8
534.525
MFCD00273454
OENCMORJQAUAAJ-DEOSSOPVSA-N
fmoc-lys dnp-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-2,4-dinitrophenyl amino hexanoic acid,fmoc-l-lys dnp-oh,n-fmoc-n'-2,4-dinitrophenyl-l-lysine,fmoc-lys dnp,2s-6-2,4-dinitrophenyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-2,4-dinitrophenyl-l-lysine
44593781
(2S)-6-(2,4-dinitroanilino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCNC4=C(C=C(C=C4)[N+](=O)[O-])[N+](=O)[O-])C(=O)O
Formula Weight 534.52
Physical Form Solid
Percent Purity 98%
Quantity 5g
Chemical Name or Material Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine

TSCA : No

Recommended Storage : Ambient temperatures