Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nepsilon-(2-Chlorobenzyloxycarbonyl)-Nalpha-Fmoc-L-lysine, 95%

Click to view available options
Quantity:
5g
25g
133970-31-7
C29H29ClN2O6
537.009
MFCD00065664
VUEYAXRHPZGZOL-SANMLTNESA-N
fmoc-lys 2-cl-z-oh,fmoc-lys 2-cl-cbz-oh,fmoc-lys 2-chloro-z-oh,n-9-fluorenylmethyloxycarbonyl-n'-2-chlorobenzyloxycarbonyl-l-lysine,fmoc-lysine 2-chloro-z-oh,fmoc-lys cl-z-oh,fmoc-l-lys 2-cl-z-oh,fmoc-n-epsilon-2-chloro-z-l-lysine,n-alpha-fmoc-n-epsilon-2-chloro-cbz-l-lysine,n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-2-chlorobenzyloxycarbonyl-l-lysine
45073227
(2S)-6-[(2-chlorophenyl)methoxycarbonylamino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
C1=CC=C(C(=C1)COC(=O)NCCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)Cl
133970-31-7
C29H29ClN2O6
537.009
MFCD00065664
VUEYAXRHPZGZOL-SANMLTNESA-N
fmoc-lys 2-cl-z-oh,fmoc-lys 2-cl-cbz-oh,fmoc-lys 2-chloro-z-oh,n-9-fluorenylmethyloxycarbonyl-n'-2-chlorobenzyloxycarbonyl-l-lysine,fmoc-lysine 2-chloro-z-oh,fmoc-lys cl-z-oh,fmoc-l-lys 2-cl-z-oh,fmoc-n-epsilon-2-chloro-z-l-lysine,n-alpha-fmoc-n-epsilon-2-chloro-cbz-l-lysine,n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-2-chlorobenzyloxycarbonyl-l-lysine
45073227
(2S)-6-[(2-chlorophenyl)methoxycarbonylamino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
C1=CC=C(C(=C1)COC(=O)NCCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)Cl
Formula Weight 537.01
Physical Form Solid
Percent Purity 95%
Quantity 5g
Chemical Name or Material Nepsilon-(2-Chlorobenzyloxycarbonyl)-Nalpha-Fmoc-L-lysine

storageNote1 : -30° to -10°C