Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nepsilon-Acetyl-Nalpha-Fmoc-L-lysine, 98%

Click to view available options
Quantity:
250mg
1g
5g
159766-56-0
C23H26N2O5
410.47
MFCD00077409
HQLBYVWJOXITAM-NRFANRHFSA-N
fmoc-lys ac-oh,fmoc-n'-acetyl-l-lysine,nalpha-fmoc-nepsilon-acetyl-l-lysine,l-lysine, n6-acetyl-n2-9h-fluoren-9-ylmethoxy carbonyl,2s-6-acetamido-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,fmoc-lys ac,fmoc-lysine ac-oh,fmoc-l-lys ac-oh,ksc005q1n,fmoc-n-epsilon-acetyl-l-lysine
7018846
(2S)-6-acetamido-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
CC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
159766-56-0
C23H26N2O5
410.47
MFCD00077409
HQLBYVWJOXITAM-NRFANRHFSA-N
fmoc-lys ac-oh,fmoc-n'-acetyl-l-lysine,nalpha-fmoc-nepsilon-acetyl-l-lysine,l-lysine, n6-acetyl-n2-9h-fluoren-9-ylmethoxy carbonyl,2s-6-acetamido-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,fmoc-lys ac,fmoc-lysine ac-oh,fmoc-l-lys ac-oh,ksc005q1n,fmoc-n-epsilon-acetyl-l-lysine
7018846
(2S)-6-acetamido-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
CC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Quantity 250mg
Formula Weight 410.46
Percent Purity 98%
Chemical Name or Material Nepsilon-Acetyl-Nalpha-Fmoc-L-lysine

TSCA : No

Recommended Storage : Store at -20°C