Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester, 95%

Click to view available options
Quantity:
1g
5g
25g
34404-36-9
C23H31N3O8
477.514
MFCD00037915
YWLICOCXPNQJPC-KRWDZBQOSA-N
boc-lys z-osu,s-2,5-dioxopyrrolidin-1-yl 6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate,boc-lys cbz-osu,2,5-dioxopyrrolidin-1-yl 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate,boc-lys z osu,boc-lys z-o su,boc-lys-z-osu,n,a-boc-n,a-z-l-lysine hydroxysuccinimide ester,n,a-z-n,a-boc-l-lysine hydroxysuccinimide ester,a-cbz-l-lysine n-hydroxysuccinimide ester
12686578
(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoate
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
34404-36-9
C23H31N3O8
477.514
MFCD00037915
YWLICOCXPNQJPC-KRWDZBQOSA-N
boc-lys z-osu,s-2,5-dioxopyrrolidin-1-yl 6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate,boc-lys cbz-osu,2,5-dioxopyrrolidin-1-yl 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate,boc-lys z osu,boc-lys z-o su,boc-lys-z-osu,n,a-boc-n,a-z-l-lysine hydroxysuccinimide ester,n,a-z-n,a-boc-l-lysine hydroxysuccinimide ester,a-cbz-l-lysine n-hydroxysuccinimide ester
12686578
(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoate
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Beilstein 1559393
Solubility Information Slightly soluble in water.
Formula Weight 477.51
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 108°C to 117°C
Quantity 5g
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester

EINECSNumber : 251-999-7

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C