Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-Nalpha-Fmoc-D-lysine, 95%

Click to view available options
Quantity:
250mg
1g
5g
110990-07-3
C29H30N2O6
502.567
MFCD00065663
KRULQRVJXQQPQH-AREMUKBSSA-N
n-fmoc-n'-cbz-d-lysine,fmoc-d-lys z-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-benzyloxy carbonyl amino hexanoic acid,ambotzfaa1673,pubchem19673,fmoc-d-lysine z-oh,n-fmoc-n-cbz-d-lysine,2r-6-benzyloxycarbonylamino-2-9h-fluoren-9-ylmethoxycarbonylamino hexanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-benzyl-oxycarbonyl-d-lysine,n~6~-benzyloxy carbonyl-n~2~-9h-fluoren-9-yl methoxy carbonyl-d-lysine
40488844
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(phenylmethoxycarbonylamino)hexanoic acid
C1=CC=C(C=C1)COC(=O)NCCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
110990-07-3
C29H30N2O6
502.567
MFCD00065663
KRULQRVJXQQPQH-AREMUKBSSA-N
n-fmoc-n'-cbz-d-lysine,fmoc-d-lys z-oh,r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-benzyloxy carbonyl amino hexanoic acid,ambotzfaa1673,pubchem19673,fmoc-d-lysine z-oh,n-fmoc-n-cbz-d-lysine,2r-6-benzyloxycarbonylamino-2-9h-fluoren-9-ylmethoxycarbonylamino hexanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-benzyl-oxycarbonyl-d-lysine,n~6~-benzyloxy carbonyl-n~2~-9h-fluoren-9-yl methoxy carbonyl-d-lysine
40488844
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(phenylmethoxycarbonylamino)hexanoic acid
C1=CC=C(C=C1)COC(=O)NCCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Formula Weight 502.57
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 106°C to 110°C
Quantity 250mg
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-Nalpha-Fmoc-D-lysine

storageNote1 : -30° to -10°C