Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nepsilon-Boc-Nalpha-Fmoc-Nepsilon-isopropyl-L-lysine, 94%

Click to view available options
Quantity:
250mg
1g
201003-48-7
C29H38N2O6
510.631
MFCD00080276
LUGFCMICCJNLBC-VWLOTQADSA-N
n-fmoc-n'-boc-n'-isopropyl-l-lysine,fmoc-lys boc isopropyl-oh,fmoc-lys ipr,boc-oh,2s-6-tert-butoxycarbonyl isopropyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,ambotzfaa1447,fmoc-l-lys boc, ipr-oh,n-9-fluorenylmethyloxycarbonyl-n'-tert-butoxycarbonyl-n'-isopropyl-l-lysine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl isopropyl amino hexanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-n-epsilon-i-propyl-l-lysine,n~6~-tert-butoxycarbonyl-n~2~-9h-fluoren-9-yl methoxy carbonyl-n~6~-propan-2-yl-l-lysine
51340513
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonyl-propan-2-ylamino]hexanoic acid
CC(C)N(CCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)OC(C)(C)C
201003-48-7
C29H38N2O6
510.631
MFCD00080276
LUGFCMICCJNLBC-VWLOTQADSA-N
n-fmoc-n'-boc-n'-isopropyl-l-lysine,fmoc-lys boc isopropyl-oh,fmoc-lys ipr,boc-oh,2s-6-tert-butoxycarbonyl isopropyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,ambotzfaa1447,fmoc-l-lys boc, ipr-oh,n-9-fluorenylmethyloxycarbonyl-n'-tert-butoxycarbonyl-n'-isopropyl-l-lysine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl isopropyl amino hexanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-n-epsilon-i-propyl-l-lysine,n~6~-tert-butoxycarbonyl-n~2~-9h-fluoren-9-yl methoxy carbonyl-n~6~-propan-2-yl-l-lysine
51340513
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonyl-propan-2-ylamino]hexanoic acid
CC(C)N(CCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)OC(C)(C)C
Formula Weight 510.63
Physical Form Solid
Percent Purity 94%
Quantity 1g
Chemical Name or Material Nepsilon-Boc-Nalpha-Fmoc-Nepsilon-isopropyl-L-lysine

storageNote1 : -30° to -10°C