Thermo Scientific NERL™ HEMA-Trol™ Whole Blood HGB Controls

Ensure accurate test results on HemoCue™ B-Hemoglobin and Hb 201+ photometers with Thermo Scientific™ NERL™ HEMA-TROL™ Whole Blood HGB Controls.

Manufacturer: Thermo Scientific 755963

 View more versions of this product

Catalog No. 22-806-003

  Check Availability
Add to Cart

    Specifications

    Stability 30 days at room temperature; 60 days at 2 to 8°C
    Description HEMA-TROL™ Normal
    For Use With (Equipment) HemoCue™ B-Hemoglobin and Hb 201+ Photometers
    Shelf Life 2 years when stored at 2 to 8°C
    Packaging 6 Pk.
    Quantity 6 x 3mL,6 x 3mL
    Level Normal,Normal
    Compatibility HemoCue™ B-Hemoglobin and Hb 201+ Photometers

    Ensure accurate test results on HemoCue™ B-Hemoglobin and Hb 201+ photometers with Thermo Scientific™ NERL™ HEMA-TROL™ Whole Blood HGB Controls. These multi-level reference controls utilize stabilized whole human red cells that process like a fresh patient sample.

    • HEMA-Trol HGB Control may be used as one would use whole blood in obtaining the stated parameters for HemoCue photometers
    • Utilizes stabilized whole human red cells which process like a fresh patient sample
    • May not be appropriate for certain instruments other than the HemoCue Photometer
    • Plastic dropper bottles for easy application

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit