Thermo Scientific™ NERL™ Reagent-Grade Water, 1 pint Available on GSA/VA Contract for Federal Government customers only.

Improve control in critical diagnostic procedures by using this reagent grade water in place of deionized or distilled water.

Catalog No. 23-769893

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »

Alert

  • For in vitro diagnostic use and laboratory use only
  • Not for infusion, injection or irrigation


    Specifications

    Certifications/Compliance Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water. Certificate of Analysis is available online.
    Quantity 1 pt.,1 pt.
    Shelf Life 12 months
    Storage Requirements 15° to 30°C

    Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ Reagent Grade Water in place of deionized or distilled water. This water meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water.

    • Ideal for reconstituting chemistry or coagulation products, preparing analytical standards or rinsing delicate electrodes
    • Prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration and final filtration to 0.2 micron
    • Essentially free from organic and inorganic particulate and soluble contaminants
    • Contains no preservatives

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit