Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nerolidol, Natural Isolate, Ricca Chemical
Nerolidol, Natural Isolate is a natural isolate food additive.
$39.70 - $979.00
Specifications
Chemical Name or Material | Nerolidol |
---|---|
CAS | 7212-44-4 |
Packaging | Amber Glass |
Color | Colorless to Light Yellow |
Certificate of Analysis | Yes |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-184-938
|
Ricca Chemical Company
RCCT905520C6 |
20 g |
Each for $39.70
|
|
|||||
01-184-918
|
Ricca Chemical Company
RCCT9055100C6 |
100 g |
Each for $144.00
|
|
|||||
01-184-979
|
Ricca Chemical Company
RCCT9055750C6 |
750 g |
Each for $979.00
|
|
|||||
Description
- All Natural
- Food Grade
- FSSC 22000 and ISO 9001 Accreditated Facility
- Additional Food Safety Testing Included (Pesticide, Residual Solvents, Heavy Metals)
Specifications
Nerolidol | |
Amber Glass | |
Yes | |
80% to 100% | |
227.37 | |
Floral, rose-like, apple-like |
7212-44-4 | |
Colorless to Light Yellow | |
Food Additive Standard | |
C15H26O | |
Food Grade | |
Liquid |
Safety and Handling
ShelfLife : 24 Months
Recommended Storage : 15°C to 30°C (59°F to 86°F)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title