Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner NERYL DIPHOSPHATE 10 mg  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner I012010MG
Isoprenoid compounds are a diverse group of natural products which are essential components in all cells. Isoprenoids are biosynthesized from the simple precursors isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP). Eukaryotes, fungi, and some gram-positive bacteria produce IPP through the mevalonate (MVA) pathway whereas gram-negative and some gram-positive bacteria utilize the non-mevalonate or 2-C-methyl-D-erythritol-4-phosphate (MEP) pathway. Neryl diphosphate is a precursor for monoterpenes such as mycrene and cyclic monoterpenes such as limonene.
Catalog No. 50-720-386