Thermo Scientific™ Temperature Control Connectivity Accessories Available on GSA/VA Contract for Federal Government customers only.
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
Manufacturer: Thermo Scientific™ 422000000004
 View more versions of this product
Catalog No. 13-265-176
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) RS-232 Communications Interface; Windows-based PC
    Type Software
    Description NEScom™ Software 4.0
    For Use With RS-232 communications interface; Windows-based PC
    Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
    • Choose upper and lower temperature alarm limits and monitor system status with an alarm
    • Record your results on a user selectable graph
    • Select from easy–to–use product icons
    • Create graphs and charts
    • Easily configure ramping functions over time
    • View pop-up alarm windows which display the error condition
    • View a virtual controller screen
    • PC compatible only
  • Description & Specifications
Related Products
Product Content Feedback