Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nesfatin-1/Nucleobindin-2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen Nesfatin-1/Nucleobindin-2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP157951
SDP
View Documents
Novus Biologicals
NBP157951
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

Nesfatin-1/Nucleobindin-2 Polyclonal specifically detects Nesfatin-1/Nucleobindin-2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Nesfatin-1/Nucleobindin-2
Polyclonal
Purified
RUO
DNA-binding protein NEFA, NEFAGastric cancer antigen Zg4, nucleobindin 2, nucleobindin-2, nucleobinding 2
NUCB2
IgG
Protein A purified
Western Blot
Unconjugated
Rabbit
P80303
4925
Synthetic peptides corresponding to NUCB2 (nucleobindin 2) The peptide sequence was selected from the C terminal of NUCB2. Peptide sequence FFTEEELKEYENIIALQENELKKKADELQKQKEELQRQHDQLEAQKLEYH.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.