Learn More
Pyramex™ Neshoba™ Safety Eyewear
Click to view available options
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor, Mirror
Lens Coating:
Scratch-resistant
Description
Specifications
Certifications/Compliance | ANSI Z87.1 |
Certifications/Compliance (Europe) | EN-166 |
Lens Tint | Gray |
Weight (Metric) | 16 g |
Lens Material | Polycarbonate |