Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nesprin2 Mouse anti-Human, Janelia Fluor 549, Clone: 6C11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259944JF549

Catalog No. NBP59944549


Add to cart

Description

Description

Nesprin2 Monoclonal antibody specifically detects Nesprin2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Nesprin2
6C11
Immunocytochemistry/Immunofluorescence
Purified
SYNE2
GST fused to residues 6,653-6,835 of human Nesp2
Protein A purified
Store at 4°C in the dark.
Monoclonal
Specificity of human Nesprin 2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunocytochemistry, Immunofluorescence
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
DKFZP434H2235, DKFZp686E01115, DKFZp686H1931, EDMD5, FLJ46790, KIAA1011FLJ11014, nesprin 2, nesprin-2, NUAFLJ43727, NUANCEFLJ45710, Nuclear envelope spectrin repeat protein 2, Nucleus and actin connecting element protein, polytrophin, Protein NUANCE, spectrin repeat containing, nuclear envelope 2, Synaptic nuclear envelope protein 2, synaptic nuclei expressed gene 2, SYNE-2, TROPH
Mouse
IgG2b Kappa
0.1mL
Primary
23224
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit