Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris™ NESS 0327
Ultra-high affinity and selective CB1 antagonist
$243.33 - $1021.11
Specifications
Molecular Formula | C24H23Cl3N4O |
---|---|
Purity | 98% |
Target | Cannabinoid R1/CB1 Antagonists |
Chemical Name or Material | 8-Chloro-1-(2,4-dichlorophenyl)-1,4,5,6-tetrahydro-N-1-piperidinyl-benzo[6,7 ]cyclohepta[1,2-c]pyrazole-3-carboxamide |
CAS | 494844-07-4 |
Description
Ultra-high affinity and selective CB1 receptor antagonist (Ki = 350 fM). Exhibits ∽60-fold selectivity for CB1 over CB2 receptors. Antagonizes the inhibitory effects of WIN 55,212-2 (Cat. No. 1038) on electrically evoked contractions in mouse isolated vas deferens ex vivo. Also antagonizes the antinociceptive effect of WIN 55,212-2 in tail-flick and hot plate tests in vivo.Specifications
C24H23Cl3N4O | |
Cannabinoid R1/CB1 Antagonists | |
494844-07-4 |
98% | |
8-Chloro-1-(2,4-dichlorophenyl)-1,4,5,6-tetrahydro-N-1-piperidinyl-benzo[6,7 ]cyclohepta[1,2-c]pyrazole-3-carboxamide |
Safety and Handling
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title