Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nestin Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA586014
Description
Rabbit Polyclonal Antibody
Nestin Polyclonal antibody specifically detects Nestin in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western BlotSpecifications
Nestin | |
Polyclonal | |
Unconjugated | |
Nes | |
AA166324; C78523; ESTM46; FLJ21841; Nbla00170; Nes; Nestin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10763, 18008 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P48681, Q6P5H2 | |
Nes | |
Synthetic peptide corresponding to Human Nestin aa 1720-1750 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title