Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Netilmicin sulfate, 595μg/mg

Click to view available options
Quantity:
10mg
25mg
56391-57-2
C21H43N5O11S
573.659
MFCD01311715
PQHZSWZPVGHDEZ-UIQTUGNFSA-N
1-N-Ethylsisomicin
23581789
(2R,3R,4R,5R)-2-[(1S,2S,3R,4S,6R)-4-amino-3-[[(2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-6-(ethylamino)-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid
CCNC1CC(C(C(C1OC2C(C(C(CO2)(C)O)NC)O)O)OC3C(CC=C(O3)CN)N)N.OS(=O)(=O)O
56391-57-2
C21H43N5O11S
573.659
MFCD01311715
PQHZSWZPVGHDEZ-UIQTUGNFSA-N
1-N-Ethylsisomicin
23581789
(2R,3R,4R,5R)-2-[(1S,2S,3R,4S,6R)-4-amino-3-[[(2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-6-(ethylamino)-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid
CCNC1CC(C(C(C1OC2C(C(C(CO2)(C)O)NC)O)O)OC3C(CC=C(O3)CN)N)N.OS(=O)(=O)O

A broad spectrum antibiotic

Applications
Netilmicin sulfate is used as a broad spectrum antibiotic active against most Gram-negative and some Gram-positive bacteria. It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studied

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Merck Index 14,6479
Solubility Information Soluble in water
Formula Weight 720.75
Physical Form Powder
Boiling Point 868°C
Color White
Quantity 25mg
Sensitivity Light Sensitive & Hygroscopic
Chemical Name or Material Netilmicin sulfate, 590μg/mg

GHS H Statement
H360-H312-H332
May damage fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : WK2286000

storageNote1 : 2° to 8°C